Perforated Baskets, Round, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
00-00368226-38 00-00368227-38
76338-064EA 1049.4 CAD
76338-064 76338-082
Perforated Baskets, Round, Marlin Steel Wire Products
Baskets Wire Baskets
Circular material handling basket with handles.
  • Natural finish
  • With four mesh openings per linear inch
  • Handles for easy gripping
  • Stacks so that parts inside the basket do not get crushed
Ideal for washing parts, parts degreasing, ultrasonic cleaning, curing, and materials handling in a variety of applications such as chemical bath, heat treating, and cryogenics.