Material Handling Baskets, Round, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
00-00368199-38 00-00368205-38
76338-068EA 847.51 CAD
76338-068 76338-070 76338-072 76338-074 76338-076 76338-078 76338-080
Material Handling Baskets, Round, Marlin Steel Wire Products
Baskets Wire Baskets
Round material handling basket with handles.
  • Natural finish
  • With 24 mesh openings per linear inch
  • Handles for easy gripping
  • Stacks so that parts inside the basket do not get crushed
Robust quality that is ideal for washing parts, cleaning, degreasing, ultrasonic cleaning, curing, and materials handling in a variety of applications such as chemical baths, heat treating, and cryogenics.