Material Handling Baskets, Rectangular, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
00-00368185-38 00-00368190-38 00-00368191-38 00-00368187-38 00-00368188-38 00-00368189-38
76338-096EA 739.95 CAD
76338-096 76338-098 76338-100 76338-102 76338-104 76338-106 76338-108
Material Handling Baskets, Rectangular, Marlin Steel Wire Products
Baskets Wire Baskets
Rectangular material handling basket with handles.
  • Natural finish
  • With four mesh openings per linear inch
  • Handles for easy gripping
  • Stacks so that parts inside the basket do not get crushed
Robust quality that is ideal for washing parts, parts degreasing, ultrasonic cleaning, curing, and materials handling in a variety of applications such as chemical bath, heat treating, and cryogenics.
Order Now
SPECIFICATIONS
Environmentally Preferable