Expanded Metal Tote Baskets, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
00-00363273-14 00-00363280-38 00-00363275-14 00-00363276-14 00-00363277-38 00-00363278-38 00-00363279-38 00-00363272-14 00-00363274-14 00-00363281-38
76338-140EA 347.16 CAD
76338-140 76338-142 76338-144 76338-146 76338-148 76338-150 76338-152 76338-154 76338-156 76338-158
Expanded Metal Tote Baskets, Marlin Steel Wire Products
Baskets Wire Baskets
Rectangular material handling basket with handles and mesh.
  • Zinc plated or natural finish
  • 0.25" or 0.50" mesh openings
  • Handles for easy gripping
  • Stacks so that parts inside the basket do not get crushed
Ideal for washing parts, parts degreasing, ultrasonic cleaning, curing, and materials handling in a variety of applications such as chemical bath, heat treating, and cryogenics.