Parts Washing Baskets, with Locking Lid, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
00-00363231-31 00-00363232-31 00-00363233-31
76338-050EA 795.51 CAD
76338-050 76338-052 76338-054
Parts Washing Baskets, with Locking Lid, Marlin Steel Wire Products
Baskets Wire Baskets
Stainless steel mesh baskets with lid that locks.
  • Electropolished finish
  • With 30 mesh openings per linear inch
Great for material handling and parts washing/ultrasonic cleaning.