Solar Cell
Supplier: Wards
PH1320EIS
470230-444EA 67.95 CAD
470230-444
Solar Cell
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Generate electricity with a solar panel.
  • Selenium voltaic solar cell mounted on a metal base
  • Accepts two 4 mm plug type cords
Demonstrate the conversion between light energy and electricity. Connect a motor via cords to the solar cell and introduce a light source to allow the solar cell to power the motor.
Ordering information: Cords not included.
Order Now
SPECIFICATIONS
Environmentally Preferable