QApp Software Packages for Sartorius Cubis® II Laboratory Balances, Sartorius
Supplier: Sartorius
Sartorius Cubis
QP2 QP99 QP3 QP4 QP1 QP2 QP3 QP1 QP4 QP99
CA76334-430EA 1430.82 CAD
CA76334-430 76376-764 76376-766 CA76333-918 76376-768 CA76334-432 76376-836 CA76333-920 76376-780 CA76334-428
QApp Software Packages for Sartorius Cubis® II Laboratory Balances, Sartorius
Software Laboratory Software
With up to 60 optional software applications clustered into QApp packages, and with continuous software development, we support your workflows now and in the future. Choose between different packages to fulfill your demands. Each package contains powerful applications or functions to enhance and individualize your balance for your workflows.
  • Thematically structured software packages for your optimized workflows
  • Seamless connectivity to diverse laboratory information systems
  • Long-term flexibility guarantees adaptation
QApp Pharma software application package (QP1): It contains applications concerning the topic compliance with pharmaceutical-relevant guidelines as 21 CFR Part 11 and USP 39, Chapter 41. The Pharma package includes applications such as user management, digital signatures, audit trail, USP minimum weight.
QApp Advanced software application package (QP2): It includes various complex weighing applications includes evaluation. This includes applications used for density determination, percentage weighing, counting, backweighing, residual dirt analysis, residue on ignition, sieve analysis, filter weighing, checkweighing, formulation, averaging, and so on.
QApp Utility software application package (QP3): It contains weighing applications and function extensions such as bootscreen, color scheme, free formula, fiber coarseness, diameter determination, air buyoancy correction, paper weight, statistics and printing of QR|bar codes.
QApp Connectivity software application package (QP4): It includes applications for data exchange, for example to Windows file server, FTPS, StarLims, and so on.
QApp software application package all inclusive (QP99): It includes four different sub-packages for compliance (pharma (QP1)), complex weighing applications (advanced (QP2)), weighing applications and helpful tools (utilities (QP3)) and connectors for data exchange (connectivity (QP4)).