EMS High Precision Rubber Grip Cutting Tweezers, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
78325-07 78325-08 78325-09 78325-10
76376-158EA 201.45 CAD
76376-158 76376-160 76376-162 76376-164
EMS High Precision Rubber Grip Cutting Tweezers, Electron Microscopy Sciences
Forceps
Soft ESD safe ergonomic cushion grips (resistivity 10⁸ to 10⁹ Ohm) provide enhanced operator comfort and reduced finger stress when handling ESD sensitive components or small static items. Tips are carbon steel.