EMS Biological Tweezers, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
78325-3SA 78325-4SA 78325-5SA 78325-7SA
76376-190EA 174.43 CAD
76376-190 76376-200 76376-222 76376-234
EMS Biological Tweezers, Electron Microscopy Sciences
Forceps
EMS biological grade tweezers are made of a superalloy that contains nickel, chromium, and molybdenum. The Swiss made tweezers are anti-acid, anti-magnetic, and have serrated handles.