VWR® T Legal-for-Trade (NTEP) Balances with Calibration Certificate
Supplier: VWR International
76378-380 76378-382 76378-384 76378-386 76378-388 76378-474
76378-380EA 7166.71 CAD
76378-380 76378-382 76378-384 76378-386 76378-388 76378-474
VWR® T Legal-for-Trade (NTEP) Balances with Calibration Certificate
Balances and Scales
The VWR® T-Series balances offer a variety of standard features, such as internal calibration, GLP procedures, adjustable ambient condition settings, 11 application modes, and multiple weighing units. The VWR® T-Series balances featuring an NTEP Certificate of Conformance to meet an organizations legal-for-trade requirements.
  • NTEP certified for Legal-for-Trade applications
  • Class I and II options
  • Internal calibration
  • 2×RS232, wireless, ethernet, and USB (A and B) communication interfaces
  • Five year warranty"
The balances include a Certificate of Calibration showing the full testing data from an ISO/IEC 17025:2017 accredited Calibration and Testing Laboratory utilizing NIST-traceable weights. These Class I (d=0.01 mg and 0.1 mg) and Class II (d=0.001 g and 0.01 g) balances offer the highest precision in legal-for-trade applications.
The 0.001 g readability models feature a removable, glass shield.
Certifications: The VWR® T-Series balances with NTEP certification which include an ISO/IEC 17025:2017 Calibration Certificate are the answer for any organization requiring legal-for-trade certification and documented unit compliance. CE marked.
Delivery information: All units supplied with switching AC adapter, weighing pan, weighing pan supports, user manual, and quick start guide. Accessories not included.