Autosampler Vials, Headspace, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
90S332346B 90S332375BMB 90S332375BMB
10061-484CS 1335.36 CAD
10061-484 10061-488 10061-490
Autosampler Vials, Headspace, DWK Life Sciences
Vials
Round bottom vials distribute the internal pressure created at high temperatures across the glass surface and are more easily handled by robotic arms that lift the vial from the tray.
  • Designed to fit most headspace autosamplers
  • Flat finish on top and bottom to maximise heating efficiency
The beveled edge 20 mm crimp seal finish features a sturdy rim that presses into the septum for a more effective seal.
Certifications: Meets or exceeds OEM specifications.