VWR® A-Series Balances with Legal-for-Trade (NTEP) and Calibration Certificate
Supplier: VWR International
76378-394 76378-396 76378-398 76458-532 76458-534 76458-536 76458-538 76458-540 76458-542 76458-544 76458-546
76378-394EA 4675.93 CAD
76378-394 76378-396 76378-398 76458-532 76458-534 76458-536 76458-538 76458-540 76458-542 76458-544 76458-546
VWR® A-Series Balances with Legal-for-Trade (NTEP) and Calibration Certificate
Balances and Scales
The VWR® A-series Balances with NTEP Certificate and Calibration Certificate provide legal-for-trade certification along with full traceability for regulatory and compliance needs.
  • Internal calibration
  • 2×RS-232, USB (A and B), and Wi-Fi communication interfaces
  • Five year warranty
  • Tool-free assembly and diassembly of the 3-door draft shield on analytical models
The VWR® A-series balances with NTEP certification and Calibration certificate are Class I and Class II certified balances featuring an NTEP Certificate of Conformance indicating the devices meet the requirements of Handbook 44. These balances also include a certificate of calibration showing the full testing data from an ISO/IEC 17025:2017 accredited Calibration and Testing Laboratory using NIST-traceable weights. Featuring internal calibration, a multitude of communication options, and built in data protections, the VWR® A-series balances provide advanced features for any application. The large, back-lit LCD display allows for easy viewing of the weighing results. Data safety is provided via the internal memory allowing for recording up to 100000 weighings. The analytical models feature a spacious 3-door draft shield that allows for tool-free assembly and disassembly of the weighing chamber. The VWR® A-series balances with NTEP certification and Certificate of Calibration are the answer for organizations demanding precision, performance and full traceability in legal-for-trade applications.
The 0.01 and 0.1 mg readability models feature a fixed 3-door draft shield (which can not be removed). The 0.001 g readability models feature a removable, glass draft shield.
Certifications: CE marked.
Ordering information: All units supplied with switching AC adapter, weighing pan, weighing pan supports, user manual, and quick start guide. Accessories not included.