Savant™ SpeedVac™ Modular Vacuum Concentrator, Thermo Scientific
Supplier: Thermo Fisher Scientific
SPD140P1-115 SPD140P2-115
76210-574EA 53878 CAD
76210-574 76210-578
Savant™ SpeedVac™ Modular Vacuum Concentrator, Thermo Scientific
Evaporators Centrifugal Evaporators
The Thermo Scientific™ Savant™ SpeedVac™ SPD140DDA vacuum concentrator is a medium-capacity vacuum concentrator with a modular design.
  • Kit
  • Medium capacity
  • Variety of rotors
  • Heated glass safety cover
  • PTFE coated aluminum chamber
The SpeedVac™ SPD140DDA is ideal for drying aggressive organic solvents, strong acids, bases, and combinatorial chemistry solvents.
Ordering information: Supplied as different packages, please see description for details of what is included in specific kits/packages.