EMS High End Medical Tweezers, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
78920-01 78920-02 78920-03 78920-04 78920-08 78920-09
76376-374EA 148 CAD
76376-374 76376-376 76376-378 76376-380 76376-382 76376-384 76376-386 76376-388 76376-390
EMS High End Medical Tweezers, Electron Microscopy Sciences
Forceps
The medical tweezers have a high precision fine tip with perfect symmetry and balance. They are made of Inox steel with an electropolish finish and dot serrated handles for a perfect grip.
  • Can be sterilized
Order Now
SPECIFICATIONS
Environmentally Preferable