Cloud Formation Apparatus
Supplier: GSC INTERNATIONAL SE
4-30101
470323-886EA 32 CAD
470323-886
Cloud Formation Apparatus
Educational Materials Earth Science Educational Materials Meteorology Learning Activities
Demonstrate the principles of cloud formation, even when there isn't a cloud in the sky, with the cloud formation apparatus.
When atmospheric pressure is lowered in the chamber and smoke particles are introduced, a nucleus for the formation of water droplets is produced. When the bulb is compressed and then suddenly released, a cloud is formed inside the flask.
Delivery information: Instructions included.