Adjustable Speed Orbital Motion Mixers
Supplier: DLAB SCIENTIFIC INC.
18900158 8031202000 18900067 18900042 18900079 8031102000 18900034 18900044 18900035 18900020 18900021 18900022 18900023 18900024 18900043
470319-880EA 5.25 CAD
470319-880 470319-882 470319-884 470319-886 470319-888 470319-912 470319-916 470319-918 470319-920 470319-922 470319-924 470319-926 470319-928 470319-930 470319-932
Adjustable Speed Orbital Motion Mixers
Shakers Vortex Mixers
Select the orbital motion mixer that best fits your needs.
  • Multiple layouts available, covering most of your lab needs
  • Additional adapters available to accommodate various tube sizes
Vortexer can be used in either touch-activated or continuous operation modes. With vacuum suction feet for body stability, your vortexer will stay put no matter what you're mixing. The robust aluminum-cast body construction and shaded pole motor makes this a reliable, affordable choice for any laboratory. Unit is supplied with a standard cap-top, but optional adapters can be purchased for plates and tubes.
Microplate Mixer has a maintenance-free brushless DC motor for a long, dependable life. Unit is supplied with a clamp for a single microplate, but optional adapter can be purchased to allow the unit to accommodate two microplates.