Graphite Contact Cylinders, PerkinElmer
Supplier: PerkinElmer
B0504036 B0504035
CA97013-420PK 3779.73 CAD
CA97013-420 CA97013-422
Graphite Contact Cylinders, PerkinElmer
Tubes Graphite Tubes
The graphite contact cylinders are engineered for a precise fit so that variations in electrical contact, which might affect the analytical performance, are reduced to a minimum.
  • Made from a proprietary blend of base graphite material
  • Produced within tight tolerances to guaranty exact fitment and maximize performance
  • Modified contacts used in applications to minimize condensation and improve analytical performance with difficult matrices (e.g., 2% Ni solutions, edible oils, etc.)
The proper performance, maintenance and serviceability of PerkinElmer graphite furnace systems can be assured only through the use of genuine PerkinElmer supplied graphite components such as these contact cylinders.
Fits PerkinElmer PinAAcle 900T, PinAAcle 900Z, AAnalyst 600, AAnalyst 800, Model 4100ZL, Model 4110ZL, Model 5100ZL, SIMAA 6000 Zeeman AA instruments equipped with a graphite furnace.