KIMBLE® Stirrer Bearings, PTFE and Glass, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
781050-2440 781050-3445
KT781050-2440EA 470.46 CAD
KT781050-2440 KT781050-3445
KIMBLE® Stirrer Bearings, PTFE and Glass, DWK Life Sciences
Stirrers Stirring Tools
These bearings are ideal for use with series KT786500-0261, KT786500-0262, KT786500-0263, KT786500-0264 glass 10 mm precision ground stirring shafts.
Threaded glass ground joint bearings seal against the stirrer shaft by means of a PTFE O-ring, a precision bore PTFE sleeve bearing and a PTFE tightening screw with lock nut and washer.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable