KIMAX® Sample Vials, Borosilicate Glass, with Unattached Black Phenolic Caps and PTFE Faced White Rubber Liners, DWK Lifesciences
Supplier: DWK Life Sciences (KIMBLE)
60912B-1 60912B-12 60912B-1232 60912B-2
89201-866CS 464.34 CAD
89201-866 89201-868 89201-870 89201-872
KIMAX® Sample Vials, Borosilicate Glass, with Unattached Black Phenolic Caps and PTFE Faced White Rubber Liners, DWK Lifesciences
Vials
Amber Vials with attached black phenolic caps and white rubber liners.
  • Closed top black phenolic closure with white rubber liners, attached
  • Manufactured from 51 expansion borosilicate amber glass conforming to UPS Type I requirement
Packaging: Packaged in corrugated trays with partitions.