Dumont Replaceable Tip Tweezers for Chemistry, Dumont Switzerland
Supplier: ELECTRON MICROSCOPY SCIENCE
76723-13 76723-14 76723-15 76723-16
76375-972EA 286.66 CAD
76375-972 76375-974 76375-976 76375-978
Dumont Replaceable Tip Tweezers for Chemistry, Dumont Switzerland
Forceps
The replaceable tip tweezers have been adapted for chemistry, radiochemistry, and radiation metrology. The tweezers are made of materials meeting the highest quality standards and feature long sleeve and delrin replaceable tips.