Dumont Flask Tongs, Dumont Switzerland
Supplier: ELECTRON MICROSCOPY SCIENCE
76723-10 76723-11 76723-12
76375-966EA 170.88 CAD
76375-966 76375-968 76375-970
Dumont Flask Tongs, Dumont Switzerland
Tongs
The tongs have a long arm that protects the user when grabing flasks used in chemistry, radiochemistry, and radiation metrology.
  • Tong like tool features a constant spring
  • Non-welded
  • One piece body
  • matte finish
Made of materials meeting the highest quality standards. They have a superior grip and hold of the object and their coated tips reduce antistatic charge.