Avantor® Partisil ODS3, HPLC Columns, 10 µm
Supplier: VWR
Partisil®
4223-001 4230-125 4682-1502 4228-001
76398-326EA 2202.5 CAD
76398-326 76382-664 76382-670 76398-244
Avantor® Partisil ODS3, HPLC Columns, 10 µm
Chromatography Columns
Avantor® Partisil® was one of the first commercially available irregular silicas. A large surface area gives it a high loading capacity.
  • Irregular porous silica
  • 5 and 10 µm particle sizes
  • Hichrom high efficiency
  • Stainless steel column hardware
Avantor® Partisil® ion-exchange materials are widely referenced and remain one of the most popular choices for analysts. Avantor® Partisil® 10 ODS3 is similar to Waters µBondapak.
It is an intermediate carbon load C18 phase used for pharmaceutical, natural products, food, biological and environmental pollutants applications.