Avantor® Partisil SCX, HPLC Columns, 10 µm
Supplier: Avantor
4227-001 4682-1507 4141-001 4222-228 4681-1507
76382-668EA 2597.5 CAD
76382-668 76382-676 76398-222 76398-230 76398-322
Avantor® Partisil SCX, HPLC Columns, 10 µm
Chromatography Columns
Avantor® Partisil® was one of the first commercially available irregular silicas. A large surface area gives it a high loading capacity.
  • Irregular porous silica
  • 5 and 10 µm particle sizes
  • Hichrom high efficiency
  • Stainless steel column hardware
Avantor® Partisil® ion-exchange materials are widely referenced and remain one of the most popular choices for analysts. Avantor® Partisil® SCX is a strong cation-exchange phase based on benzenesulphonic acid groups
Ideal for nucleosides, amino acids, polyamines, drugs and other cationic species.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001:2015 (530840 UM15)