Avantor® ACE® Guard Column Cartridge Holders
Supplier: Avantor
H0001 H0002 H0009 H0011 BIO-GRD-0001
76382-654EA 226.26 CAD
76382-654 76382-656 76395-216 76395-220 76395-228 76395-232 76457-990
Avantor® ACE® Guard Column Cartridge Holders
Guard Column Holders
Stainless steel Avantor® ACE® stand-alone and integral guard cartridge holders are used for connecting guard cartridges to the LC column to provide column protection.
Integral holders connect directly to the analytical column. Stand-alone holders require the use of a suitable connector.