Avantor® ACE® Excel® C18-PFP, Analytical UHPLC Columns, 5 µm
Supplier: Avantor
EXL-1210-1546U EXL-1210-0502U EXL-1210-0503U EXL-1210-0546U EXL-1210-1002U EXL-1210-1003U EXL-1210-1046U EXL-1210-1502U EXL-1210-2502U EXL-1210-2503U EXL-1210-2546U
76382-646EA 2062.96 CAD
76382-646 76393-742 76393-744 76393-746 76393-748 76393-750 76393-752 76393-754 76393-756 76393-758 76393-760 76393-762 76393-764 76393-766 76393-768 76393-770 76393-772 76393-774 76393-782 76393-784 76393-786 76393-794 76393-796 76393-798 76393-806 76393-808 76393-810
Avantor® ACE® Excel® C18-PFP, Analytical UHPLC Columns, 5 µm
Chromatography Columns
Avantor® ACE® Excel® C18-PFP phase utilizes a specially developed ligand combining a C18 chain with integral PFP functionality. This results in a phase that maintains the hydrophobic, stability and low bleed characteristics of leading C18 phases, yet provides the multiple retention mechanisms of a PFP phase.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1.7 to 5 µm) and complementary phases
  • Also useful for analytes with π-bonding or electron or proton donor groups
  • Ultra-low bleed to ensure LC/MS compatibility
  • Method validation kits contain three different batches of the same phase - available on request
Due to its integral pentafluoro phenyl functionality, Avantor® ACE® C18-PFP is also recommended for separations that involve halogenated aromatic compounds, Regio isomers, and those analytes with differing shape constraints.
These columns are made of stainless steel.
Ordering information: Method development and validation kits are available on request. Please contact chromsupport@avantorsciencs.com.