Avantor® ACE® Excel® C18-PFP, Analytical UHPLC Columns, 1.7 µm
Supplier: Avantor
EXL-1710-1002U EXL-1710-0202U EXL-1710-0302U EXL-1710-0502U EXL-1710-0503U EXL-1710-1003U EXL-1710-3502U EXL-1710-7502U EXL-1710-7503U
76382-648EA 3137.3 CAD
76382-648 76394-404 76394-406 76394-408 76394-410 76394-412 76394-414 76394-416 76394-428 76394-430 76394-436 76394-438
Avantor® ACE® Excel® C18-PFP, Analytical UHPLC Columns, 1.7 µm
Chromatography Columns
Avantor® ACE® Excel® C18-PFP phase utilizes a specially developed ligand combining a C18 chain with integral PFP functionality. This results in a phase that maintains the hydrophobic, stability and low bleed characteristics of leading C18 phases, yet provides the multiple retention mechanisms of a PFP phase.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1.7 to 5 µm) and complementary phases
  • Also useful for analytes with π-bonding or electron or proton donor groups
  • Ultra-low bleed to ensure LC/MS compatibility
  • Suitable for use with 100% aqueous mobile phases
  • Method validation kits contain three different batches of the same phase - available on request
Due to its integral pentafluoro phenyl functionality, Avantor® ACE® C18-PFP is also recommended for separations that involve halogenated aromatic compounds, Regio isomers, and those analytes with differing shape constraints.
These columns are made from stainless steel.