Avantor® ACE® Excel® C4, Analytical HPLC/UHPLC Columns, 2 µm
Supplier: Avantor
EXL-103-1002U EXL-103-0502U EXL-103-1502U EXL-103-1503U EXL-103-3503U EXL-103-7502U EXL-103-7503U ACE-1P3-GD2U ACE-1P3-GD4U
76382-620EA 2731.52 CAD
76382-620 76392-756 76392-758 76392-760 76392-762 76392-764 76392-766 76392-768 76392-770 76392-772 76392-774 76392-776 76392-778 76392-780 76392-782 76392-784 76392-786 76392-788 76392-790 76392-792 76392-794 76392-796 76392-798 76392-800 76408-418 76408-420
Avantor® ACE® Excel® C4, Analytical HPLC/UHPLC Columns, 2 µm
Chromatography Columns
These ultra-inert Avantor® ACE® Excel® C4 columns provide great reproducibility and column lifetime. These columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 5 µm) and complementary phases available
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.