Q-Switch/Alex LaserShields, 532/755/1064 nm, NoIR Laser Company
Supplier: NOIR MEDICAL
TRI52 TRI53 TRI60
76332-546EA 562.99 CAD
76332-546 76332-548 76332-550
Q-Switch/Alex LaserShields, 532/755/1064 nm, NoIR Laser Company
Safety Glasses
CE and ANSI certified laser protection eyewear.
  • Lightweight polycarbonate
Available in sporty frame #52, fit-over frame #53, or goggle frame #60.
Certifications: CE Certified LaserShields meet ANSI Z136.1, EN 207/EN 208/EN 60825, and Z87.1 for impact.
Ordering information: Hard-shell case, cleaning cloth, and strap included.