Ytterbium (YB), 1030 to 1064 nm LaserShields, NoIR Laser Company
Supplier: NOIR MEDICAL
YG452 YG453 YG460
76332-540EA 586.42 CAD
76332-540 76332-542 76332-544
Ytterbium (YB), 1030 to 1064 nm LaserShields, NoIR Laser Company
Safety Glasses
CE and ANSI certified laser protection eyewear.
  • Lightweight polycarbonate
Available in sporty frame #52, fit-over frame #53, or goggle frame #60.
Certifications: CE Certified LaserShields meet ANSI Z136.1, EN 207/EN 208/EN 60825, and Z87.1 for impact.
Ordering information: Hard-shell case, cleaning cloth, and strap included.