Holmium LaserShields, NoIR Laser Company
Supplier: NOIR MEDICAL
HOY52 HOY53 HOY60
76332-484EA 410.53 CAD
76332-484 76332-486 76335-170
Holmium LaserShields, NoIR Laser Company
Safety Glasses
CE and ANSI certified laser protection eyewear.
  • Glass lenses
Available in sporty frame #46, fit-over frame #52, or goggle frame #60.
Certifications: CE Certified LaserShields meet ANSI Z136.1, EN 207/EN 208/EN 60825, and Z87.1 for impact.
Ordering information: Hard-shell case, cleaning cloth, and strap included.