CO₂ Laser/Eximer LaserShields, NoIR Laser Company
Supplier: NOIR MEDICAL
EC252 EC253
76332-472EA 222.87 CAD
76332-472 76332-474
CO₂ Laser/Eximer LaserShields, NoIR Laser Company
Safety Glasses
CE and ANSI certified laser protection eyewear.
  • Lightweight polycarbonate
Available in sporty frame #52, fit-over frame #53, or goggle frame #60.
Certifications: CE Certified LaserShields meet ANSI Z136.1, EN 207/EN 208/EN 60825, and Z87.1 for impact.
Ordering information: Hard-shell case, cleaning cloth, and strap included.