Avantor® Hichrom, PEEK Finger Tight Fittings
Supplier: Avantor
HI-031 HI-050X HI-047 HI-050 HI-058
76382-678EA 194.86 CAD
76382-678 76382-680 76399-022 76399-024 76399-028
Avantor® Hichrom, PEEK Finger Tight Fittings
Fittings and Connectors
Universal fitting, suitable for most HPLC connections. Unlike stainless steel fittings, PEEK finger tight fittings will not permanently lock into place on the tubing. Compatible with all types of end fittings.
  • Finger tight to 5000 psi
  • Void-free connection
  • Inert and biocompatible
  • Excellent chemical resistance
Can be used at temperatures up to 150 °C and with stainless steel and PEEK tubing.