Avantor® ACE® Phenyl Ultra-Inert, HPLC Columns, 3 µm
Supplier: VWR
Avantor® ACE®
ACE-215-0501S ACE-215-3546 ACE-215-1001S ACE-215-15001 ACE-215-0103GD ACE-215-1201 ACE-215-3504 ACE-215-05005 ACE-215-0301 ACE-215-3502 ACE-215-12003 ACE-215-0546 ACE-215-12005S ACE-215-0203 ACE-215-10001 ACE-215-0101GD ACE-215-0502 ACE-215-1203 ACE-215-1502 ACE-215-7503 ACE-215-0102GD ACE-215-0302 ACE-215-7546 ACE-215-25003 ACE-215-1246 ACE215005005GD ACE-215-1501S ACE-215-35005 ACE-215-3501 ACE-215-1546 ACE-215-1001 ACE-2151000075 ACE-215-1503 ACE-215-3503 ACE-215-05005S ACE-215-0503 ACE-215-15005S ACE-215-05003 ACE-2151500075 ACE-215-10005 ACE-215-7501S ACE-215-75005 ACE-215-1501 ACE-215-0301S ACE-215-1004 ACE-215-25005 ACE-215-0501 ACE-215-0303 ACE-215-1003 ACE-215-75003 ACE-215-1201S ACE215005003GD ACE-215-03003 ACE-215-1202 ACE-215-0504 ACE-215-10003 ACE-215-7501 ACE-215-10005S ACE-215-0246 ACE-215-7504 ACE-215-1504 ACE-215-75005S ACE-215-0202 ACE-215-1002 ACE-215-1046 ACE-215-15005 ACE-215-03005 ACE-215-15003 ACE-215-1204 ACE-215-0346 ACE-215-03005S ACE-215-7502 ACE-215-12005
76390-110EA 1521.1 CAD
76390-110 76390-034 76390-078 76390-076 76390-032 76390-030 76390-074 76382-594 76390-072 76390-070 76390-118 76390-116 76390-114 76390-038 76390-112 76390-036 76390-044 76390-088 76390-122 76390-042 76390-086 76390-120 76390-040 76390-084 76390-082 76390-080 76390-008 76390-006 76390-128 76390-048 76390-004 76390-126 76390-046 76390-124 76390-132 76390-012 76390-056 76390-130 76390-010 76390-054 76390-098 76390-096 76390-052 76390-094 76390-050 76390-092 76390-090 76390-138 76390-018 76390-136 76390-016 76390-134 76390-058 76390-014 76390-066 76390-022 76390-144 76390-100 76390-064 76390-020 76390-142 76390-062 76390-140 76390-060 76390-108 76390-028 76390-106 76390-026 76390-104 76390-068 76390-024 76390-146 76390-102
Avantor® ACE® Phenyl Ultra-Inert, HPLC Columns, 3 µm
Chromatography Columns
These ultra inert Avantor® ACE® Phenyl-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
  • Designed for peptide and protein separations
  • Excellent peak shape even at extremely low levels of additives such as TFA, making them ideal for LC/MS
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.