Avantor® ACE® AQ, HPLC Columns, 3 µm
Supplier: Avantor
ACE-116-1503 ACE-116-0101GD ACE-116-0103GD ACE-116-0202 ACE-116-03005 ACE-116-0301 ACE-116-0302 ACE-116-0501 ACE-116-0502 ACE-116-0503 ACE-116-0546 ACE-116-10005 ACE-116-1002 ACE-116-1003 ACE-116-1004 ACE-116-1046 ACE-116-1202 ACE-116-1204 ACE-116-1501 ACE-116-1502 ACE-116-1504 ACE-116-1546 ACE-116-3502 ACE-116-7502 ACE-116-7503
76382-564EA 2589.56 CAD
76382-564 76385-200 76385-202 76385-204 76385-206 76385-208 76385-210 76385-212 76385-214 76385-216 76385-218 76385-220 76385-222 76385-224 76385-226 76385-228 76385-230 76385-232 76385-234 76385-236 76385-238 76385-240 76385-242 76385-244 76385-246 76385-248 76385-250 76385-252 76385-254 76385-256 76385-258 76385-260 76385-262 76385-264 76385-266 76385-268 76385-270 76385-272 76385-274 76385-276 76385-278 76385-280 76385-282 76385-284 76385-286 76385-288 76385-290 76385-292 76385-294 76385-296 76385-298 76385-300 76385-302 76385-304 76385-306 76385-308 76385-310 76385-316 76385-320 76385-322 76385-324 76385-326 76385-328 76385-330 76385-332 76385-334 76385-336 76385-338 76385-340 76385-342 76385-344 76385-346 76385-348
Avantor® ACE® AQ, HPLC Columns, 3 µm
Chromatography Columns
Avantor® ACE® AQ columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Retains polar compounds in 100% aqueous mobile phase
  • Unique C18 bonded phase with integral polar functionality
  • Ideal for use with fast gradients due to its rapid re-equilibration properties and resistance to retention loss
  • Wide range of particle sizes (3 to 10 µm) and complementary
More Product Information