Avantor® ACE® Phenyl, HPLC Columns, 3 µm
Supplier: Avantor
ACE-115-0502 ACE-115-1546 ACE-115-0102GD ACE-115-0103GD ACE-115-7546 ACE-115-05003 ACE-115-0503 ACE-115-0546 ACE-115-1002 ACE-115-1003 ACE-115-1046 ACE-115-1502 ACE-115-1503 ACE-115-1504 ACE-115-1539
76382-560EA 2305.57 CAD
76382-560 76382-562 76385-042 76385-044 76385-046 76385-048 76385-050 76385-052 76385-054 76385-056 76385-058 76385-060 76385-062 76385-196 76385-198 76385-064 76385-066 76385-068 76385-070 76385-072 76385-074 76385-076 76385-078 76385-080 76385-082 76385-084 76385-086 76385-088 76385-090 76385-092 76385-094 76385-096 76385-098 76385-100 76385-102 76385-104 76385-106 76385-108 76385-110 76385-112 76385-114 76385-116 76385-118 76385-120 76385-122 76385-124 76385-126 76385-128 76385-130 76385-132 76385-134 76385-136 76385-138 76385-140 76385-142 76385-144 76385-146 76385-148 76385-150 76385-152 76385-154 76385-156 76385-158 76385-160 76385-166 76385-170 76385-172 76385-174 76385-176 76385-178 76385-180 76385-182 76385-184 76385-186 76385-188 76385-190 76385-192 76385-194
Avantor® ACE® Phenyl, HPLC Columns, 3 µm
Chromatography Columns
These ultra-inert Avantor® ACE® Phenyl columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Offers strong π-π interactions
  • Ideal for analytes with π-bonding, electron withdrawing or delocalized groups
  • Wide range of particle sizes (2, 3, 5 and 10 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.