KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
747540-0520 747540-1020 747540-2520
KT747540-0520EA 48.67 CAD
KT747540-0520 KT747540-1020 KT747540-1520 KT747540-2520
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
Flasks Pear Shaped Flasks
These flasks have short necks for easy retrieval of residues or reaction products.
  • Accessory for Microscale MICROFLEX® threaded kits
  • Thread size: 20-400
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable