Cubis® II Premium Precision Balances, MCE Series, Automatic Leveling, Sartorius
Supplier: Sartorius
MCE4202S0-S00 MCE6202S0-S00 MCE8201S0-S00 MCE2202S0-S00 MCE8202S2S000 MCE6202S2S000 MCE6202P2S000 MCE4202S2S000 MCE2202S2S000 MCE1202S2S000 MCE12201S2S000 MCE8201S2S000 MCE5201S2S000
CA76333-820EA 7489.06 CAD
CA76333-820 CA76333-822 CA76333-826 CA76333-944 CA76334-384 CA76334-386 CA76334-388 CA76334-390 CA76334-392 CA76334-394 CA76334-396 CA76334-398 CA76334-400
Cubis® II Premium Precision Balances, MCE Series, Automatic Leveling, Sartorius
Balances and Scales
These MCE series essential user interface balance was a large, high-contrast touch display with factory-installed essential weighing applications for easy operation.
  • Error-free operation with individual QApp workflows, motorised automatic leveling
  • Full end-to-end data integrity
  • Outstanding service support integrated status center, integrated service functions preventive maintenance on accredited standards
  • User Management provides complete access control and configured in accordance with password policy
  • Gesture control allows user to open and close the automatic draft shield, start the ionizer, or send a document
  • Built-in ionizer for electrostatic elimination- electrostatic charges on samples and vessels are removed effectively
  • Maximum capacity between 320 and 14200 g scale interval with a range of 1 to 100 mg
  • Easy to use manual glass draft shield
  • Removable flat stainless-steel draft shield frame
Climate monitoring ensures that temperature, pressure, and humidity do not negatively impact results. Status center notifications show all information about balance and environmental conditions like calibration, leveling, temperature, humidity, air pressure and service, centralized on a dashboard.
It can automatically execute time-controlled actions for backup. The system is able to upload data on a file share or export the data to other systems. Safe data transfer saves all files, together with a calculated MD5 checksum which is stored in the audit trail files. Audit trail and alibi memory has a tamper-protected time-stamped electronic logfile, that allows reconstruction of events related to the creation, modification, and or deletion of records and the system automatically stores weighing data in a ring buffer.
The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis® II balance to all weighing tasks. The Cubis® II range of premium laboratory balances with a maximum load between 2.1 g and 70 kg provides the ideal model for every application. It supports automatic time synchronization through Network Time Protocol (NTP). An electronic signature can be used to sign the final report for weighing processes.
Display: State of the art TFT touch screen operation with brilliant, readable, display but uncomplex, easy to operate user interface; Software: Factory installed basic set of essential weighing applications; Operation: Activated by touch key, touch-free using IR sensor (draft shield M) or gesture sensor (optional), learning capability.
Flat, stainless steel weight pan with no draft shield for weighing modules.
Certifications: These balances ensure safe and accurate data storage and supports all requirements for FDA directive 21 CFR part 11 and EU Annex 11 compliance.
Delivery information: There are different optional software packages, each contains applications or functions to enhance and customize the balance for the user's workflows. Choice of QApp package: Pharma; advanced applications: utilities; Connectivity or the all inclusive QApps package includes all of these four packages. (denoted by QP99).