Cubis® II Essential Premium Precision Balances, Manual Leveling, Sartorius
Supplier: Sartorius
MCE12201S0-S00 MCE32202P0-S00 MCE32202P2S000 MCE14202S2S000 MCE14202P2S000 MCE10202S2S000
CA76334-150EA 7658.95 CAD
CA76334-150 CA76334-158 CA76334-376 CA76334-378 CA76334-380 CA76334-382
Cubis® II Essential Premium Precision Balances, Manual Leveling, Sartorius
Balances and Scales
These MCE series essential user interface balance was a large, high-contrast touch display with factory-installed essential weighing applications for easy operation.
  • Error-free operation with individual QApp workflows, manual leveling
  • Full end-to-end data integrity
  • Outstanding service support integrated status center, integrated service functions preventive maintenance on accredited standards
  • User Management provides complete access control and configured in accordance with password policy
  • Gesture control allows user to open and close the automatic draft
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Voltage 100 - 240 V