KIMBLE® Precision Polished Stirrer Shafts, Glass, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
786520-0260 786520-0261
46000-214EA 224.62 CAD
46000-214 46000-216
KIMBLE® Precision Polished Stirrer Shafts, Glass, DWK Life Sciences
Stirrers Stirring Shafts
Glass stirrer shaft with a precision-polished surface.
  • Shaft OD: 10 mm
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Confirms to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable