Cubis® II Essential Premium Ultra Micro Balances, MCA Series, Standard Versions, Sartorius
Supplier: Sartorius
MCA2.7S2S00F MCA2.7S2S00M MCA2.7S-M-MTS
CA76334-218EA 66662.57 CAD
CA76334-218 CA76334-226 CA76377-396 CA76377-398
Cubis® II Essential Premium Ultra Micro Balances, MCA Series, Standard Versions, Sartorius
Balances and Scales
These Cubis® II laboratory balances are ideal for high-performance portfolio of both lab weighing hardware and software.
  • Individually configured from different components
  • Fully customizable hardware, software, and connectivity
  • Customizable modularity, intelligence by design, leading Performance, high speed accuracy, error-free operations, full end to end data intergrity, reliable support
  • Automatic glass draft shield with or without integrated ionizer to eliminate interfering electrostatic charges on samples and sample containers
  • Removable flat stainless steel draft shield frame
  • Large 7" color touch display including touch, scroll, swipe functionality
  • Diverse QApp packages for optional software extension
  • Automatic, motorized leveling, gesture control, Climate monitoring, built-in ionizer for electrostatic elimination
These balances offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
The status center shows all information about your balance and environmental conditions. Automatically execute time-controlled actions for backup.
The Cubis® II balance contains an 'Alibi Memory'. The system automatically stores weighing data in a ring buffer. An electronic signature can be used to sign the final report for weighing processes. Based on a secure password, the combination of username and password is fully compliant with 21CFR.
Different draft shields and sereis are:
F - Filter draft shield micro balances
M - Motorized draft shield micro balances.
Certifications: All requirements for FDA directive 21 CFR part 11 and EU Annex 11 compliance.
Delivery information: There are different optional software packages, each contains applications or functions to enhance and customise the balance for the users’ workflows. Choice of QApp Package: Pharma; Advanced Applications: Utilities; Connectivity or the all inclusive QApps package includes all of these four packages. (denoted by QP99).