Cubis® II Essential Premium Ultra Micro Balances, MCE Series, Standard Versions, Sartorius
Supplier: Sartorius
MCE2.7S2S00M MCE2.7S2S00F
CA76334-108EA 63625.55 CAD
CA76334-108 CA76334-272
Cubis® II Essential Premium Ultra Micro Balances, MCE Series, Standard Versions, Sartorius
Balances and Scales
These Cubis® II premium MCE series - essential user interface balances are ideal for high-performance portfolio of both labs weighing hardware and software.
  • Individually configured from different components
  • Fully customizable hardware, software, and connectivity
  • Customizable modularity, intelligence by design, leading Performance, high speed accuracy, error-free operations, full end to end data integrity, reliable support
  • Easy to use manual glass draft shield, safety data transfer
  • Removable flat stainless-steel draft shield frame
  • Large 7" color touch
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Voltage 100 - 240 V