WHEATON® Diazomethane Generator, DWK Life Sciences
Supplier: DWK Life Sciences
281155
21518-134CS 385.49 CAD
21518-134
WHEATON® Diazomethane Generator, DWK Life Sciences
Gas Generators Compressed Air Stations
This Diazomethane Generator is used to generate diazomethane and other gases for gas chromatography, mass spectroscopy and NMR studies using one millimole of Methylnitronitrosoguanidine (MNNG).
  • Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to ASTM E 438 Type I, Class A requirements
Includes an “O” ring clamp, a 13-425 screw cap with a cut-out top and a PTFE faced rubber liner.
Caution: Please use a safety shield and efficient fume hood when generating diazomethane or similar gases.