Cubis® II Essential Analytical Laboratory Balances, MCE Series, Standard Version, Sartorius
Supplier: Sartorius
MCE124S-2S00-A MCE224S-U-MTS MCE524S-2S00-U MCE524P-2S00-U MCE324S-2S00-U MCE324P-2S00-U MCE224S-2S00-U MCE124S-2S00-U MCE524S-2S00-A MCE524P-2S00-A MCE324S-2S00-A MCE324P-2S00-A MCE224S-2S00-A MCE524P-2S00-I MCE324S-2S00-I MCE324P-2S00-I MCE224S-2S00-I MCE124S-2S00-I MCE524S-2S00-I
CA76333-814EA 13385.23 CAD
CA76333-814 CA76333-862 CA76333-948 CA76334-152 CA76334-276 CA76334-278 CA76334-280 CA76334-282 CA76334-284 CA76334-286 CA76334-296 CA76334-298 CA76334-300 CA76334-302 CA76334-304 CA76334-306 CA76334-308 CA76334-310 CA76334-312 CA76334-314 CA76377-298
Cubis® II Essential Analytical Laboratory Balances, MCE Series, Standard Version, Sartorius
Balances and Scales
These Cubis® II laboratory balances are ideal for high-performance portfolio of both labs weighing hardware and software.
  • Individually configured from different components
  • Fully customizable hardware, software, and connectivity
  • Customizable modularity, intelligence by design, leading Performance, high speed accuracy, error-free operations, full end to end data integrity, reliable support
  • Automatic glass draft shield with or without integrated ionizer to eliminate interfering electrostatic charges on samples and sample containers
  • Removable flat stainless-
More Product Information