Argos Technologies® Omega® Multichannel Adjustable-Volume Pipettors, Cole Parmer
Supplier: Antyila Scientific
24501-19 24501-20 24501-21 24501-22 24501-23 24501-24
76379-884EA 811.98 CAD
76379-884 76379-886 76379-888 76379-890 76379-892 76379-894
Argos Technologies® Omega® Multichannel Adjustable-Volume Pipettors, Cole Parmer
Pipettes Micropipettes
Ergonomic finger grip for comfortable use.
  • Features click-stop volume settings for simple and quick use
  • Ergonomic, lightweight push button design makes them easy to operate
  • Forward and reverse pipetting modes
  • Drop of the pipette is color-coded by volume for easy identification
  • Volume displayed through handgrip window for quick glance convenience
These pipettes are offered in a wide-range of adjustable volumes to lend to a variety of precise liquid dispensing needs. They are fully autoclavable.