Argos Technologies® Omega® Single-Channel Adjustable-Volume Pipettors, Cole Parmer
Supplier: Antyila Scientific
24501-01 24501-02 24501-03 24501-05 24501-07 24501-08 24501-09 24501-10
76379-864EA 306.59 CAD
76379-864 76379-866 76379-868 76379-870 76379-872 76379-874 76379-876 76379-878
Argos Technologies® Omega® Single-Channel Adjustable-Volume Pipettors, Cole Parmer
Pipettes Micropipettes
Ergonomic finger grip for comfortable use.
  • Easy to operate with its ergonomic, lightweight push-button design
  • Forward and reverse pipetting modes
  • Drop of the pipette is color-coded by volume for easy identification
  • Volume is displayed through handgrip window for quick glance convenience
  • Adjustable-volume pipettors feature click-stop volume settings
These pipettors are offered in a wide-range of adjustable volumes to lend to a variety of precise liquid dispensing needs. They are fully autoclavable.