Cubis® II Advanced Premium Semi Micro Balances, MCA Series, Standard Versions, Sartorius
Supplier: Sartorius
MCA225S2S00U MCA225P2S00U MCA125S2S00U MCA125P2S00U MCA225S2S00A MCA225P2S00A MCA125S2S00A MCA125P2S00A MCA225S2S00I MCA225P2S00I MCA125S2S00I MCA125P2S00I MCA125S-I-MTS
CA76333-958EA 23122.49 CAD
CA76333-958 CA76333-960 CA76333-962 CA76333-964 CA76333-978 CA76333-980 CA76333-982 CA76333-984 CA76333-998 CA76334-000 CA76334-002 CA76334-004 CA76377-336 CA76377-344 CA76377-352
Cubis® II Advanced Premium Semi Micro Balances, MCA Series, Standard Versions, Sartorius
Balances and Scales
These Cubis® II laboratory balances are ideal for high-performance portfolio of both lab weighing hardware and software.
  • Individually configured from different components
  • Fully customizable hardware, software, and connectivity
  • Customizable modularity, intelligence by design, leading Performance, high speed accuracy, error-free operations, full end to end data intergrity, reliable support
  • Automatic glass draft shield with or without integrated ionizer to eliminate interfering electrostatic charges on samples and sample containers
  • Removable flat stainless
More Product Information