DURAN® Silicone Seal, PTFE Coated (VMQ), DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
292480805 292481107 292481304 292481904 292483005
76264-258PK 19.47 CAD
76264-258 76264-260 76264-262 76264-266 76264-276
DURAN® Silicone Seal, PTFE Coated (VMQ), DWK Life Sciences
Septa
These seals are suitable for PBT screw caps and tamper-evident screw caps.
  • Autoclavable
  • Peroxidically-cured silicon
  • Heat resistance 130 °C (vapor) and 200 °C (dry heat)
Good chemical resistance due to PTFE coating.