Cubis® II Premium High Capacity Laboratory Balances, Sartorius
Supplier: Sartorius
MCA20201S0-S00 MCA32202P0-S00 MCA36201P2S000 MCA20201S2S000 MCA11201S2S000 MCA70200S2S000 MCA36200S2S000 MCA32202P2S000 MCA70201S0-S00 MCA70201S2S000 MCA50201S2S000 MCA36201S2S000 MCA36201S-0-MTS
CA76333-800EA 14610.94 CAD
CA76333-800 CA76333-812 CA76333-850 CA76333-852 CA76333-854 CA76333-856 CA76333-858 CA76334-096 CA76334-144 CA76334-258 CA76334-260 CA76334-262 CA76377-198
Cubis® II Premium High Capacity Laboratory Balances, Sartorius
These Cubis® II premium high capacity laboratory balances are ideal for high-performance portfolio of both labs weighing hardware and software.
  • Individually configured from different components
  • Fully customizable hardware, software, and connectivity
  • Customizable modularity, intelligence by design, leading Performance, high speed accuracy, error-free operations, full end to end data integrity, reliable support
  • Easy to use manual glass draft shield, safety data transfer
  • Removable flat stainless-steel draft shield frame
  • Large 7" color touch display including touch, scroll, swipe functionality
  • Diverse QApp packages for optional software extension
  • Automatic, motorized leveling, gesture control, climate monitoring, built-in ionizer for electrostatic elimination
These balances offer modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
The status center shows all information about your balance and environmental conditions. Automatically execute time-controlled actions for backup.
The Cubis® II balance contains an 'Alibi Memory'. The system automatically stores weighing data in a ring buffer. An electronic signature can be used to sign the final report for weighing processes. Based on a secure password, the combination of username and password is fully compliant with 21CFR.
Flat, stainless steel weight pan with no draft shield for weighing modules.
Certifications: All requirements for FDA directive 21 CFR part 11 and EU Annex 11 compliance.
Delivery information: There are different optional software packages, each contains applications or functions to enhance and customize the balance for the user's workflows. choice of QApp package: pharma; advanced applications: utilities; connectivity or the all-inclusive QApps package includes all of these four packages. (denoted by QP99).