KIMBLE® Inlet Gas Adapters with Septum, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
211275-1420 211275-2440
KT211275-1420EA 403.72 CAD
KT211275-1420 KT211275-2440
KIMBLE® Inlet Gas Adapters with Septum, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Inlet gas adapters have a standard taper joint.
  • Manufactured from 33 expansion, low extractable borosilicate glass
Sidearm has a 2 mm bore PTFE stopcock with a port for gas introduction and a rubber-stoppered port on the top tube for syringe retrieval.
Delivery information: Supplied with KT821001-0002 PTFE stopcock and KT882311-0000 septum stopper.