Cubis® II Essential Premium Semi-Micro Balances, MCE Series, Standard Versions, Sartorius
Supplier: Sartorius
MCE225S2S00U MCE225P2S00U MCE125S2S00U MCE125P2S00U MCE225S2S00A MCE225P2S00A MCE125S2S00A MCE125P2S00A MCE125P2S00I MCE125S2S00I MCE225P2S00I MCE225S2S00I
CA76333-940EA 16016.83 CAD
CA76333-940 CA76333-950 CA76334-116 CA76334-118 CA76334-120 CA76334-122 CA76334-288 CA76334-290 CA76334-292 CA76334-294 CA76377-224 CA76377-294 CA76377-326 CA76377-348
Cubis® II Essential Premium Semi-Micro Balances, MCE Series, Standard Versions, Sartorius
Balances and Scales
These MCE series advanced user interface balances have fully customizable hardware, software, and connectivity including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QApp packages for optional software extension.
  • Quick and easy, motorized, auto-levelling function, integrated climate sensors, individual sample holders
  • Outstanding service support integrated status center, integrated service functions preventive maintenance on accredited standards
  • User Management provides complete access control and configured in accordance with password policy
  • For all models up to a maximum capacity of 8.2 kg
Units provide modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
Audit trail and alibi memory has a tamper-protected time-stamped electronic logfile, that allows reconstruction of events related to the creation, modification, and or deletion of records and the system automatically stores weighing data in a ring buffer. The data can be displayed, filtered, and exported in an easy way. It can automatically execute time-controlled actions for backup. The system was able to upload data on a file share or export the data to other systems. Safe data transfer saves all files, together with a calculated MD5 checksum which is stored in the audit trail files. An electronic record should be protected against any manipulation.
The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis® II balance to all weighing tasks. The Cubis® II range of premium laboratory balances with a maximum load between 2.1 g and 70 kg provides the ideal model for every application. It supports automatic time synchronization through Network Time Protocol (NTP). An electronic signature can be used to sign the final report for weighing processes.
This balance has a state of the art TFT touch screen operation with brilliant, readable, display but uncomplex, easy to operate user interface. Consists of software factory installed basic set of essential weighing applications. Operations are activated by touch key, touch-free using IR sensor (draft shield M) or gesture sensor (optional), learning capability. Maximum capacity ranges between 120 g and 520 g, scale interval with a range of 0.01 to 0.1 mg. Automatic glass draft shield with or without integrated ionizer to eliminate interfering electrostatic charges on samples and sample containers. Manual glass draft shield with smooth-action doors.
Draft shield: A- motorized draft shield; I - motorized draft shield + ionizer; U - manual glass draft shield.
Certifications: The Cubis® II balances ensure safe and accurate data storage and support all requirements for FDA directive 21 CFR Part 11 compliance and EU Annex 11 compliance.
Delivery information: There are different optional software packages, each contains applications or functions to enhance and customize the balance for the user's workflows. Choice of QApp package: Pharma; advanced applications: utilities; connectivity or the all inclusive QApps package includes all of these four packages. (denoted by QP99).